Key Documents
Z137219
Ace round-bottom flask with threaded side-arm
capacity 250 mL
Synonym(s):
bottom flask, flask round, round bottom flasks
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
short
two-neck
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Ace Glass 6936-35
capacity
250 mL
General description
24/40 joint. Side-arm is internally threaded for use with a nylon bushing. Will accommodate thermometers or bleed tubes up to 7 mm diam. Complete with bushing and FETFE® O-ring.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.