Skip to Content
Merck
All Photos(1)

Key Documents

917265

Sigma-Aldrich

Tapered dispensing tips

22G, 0.50″, 50/box

Synonym(s):

3d bioprinting consumables, Bioprinting nozzle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42182011
NACRES:
NB.51

Quality Level

description

Tapered dispensing tips, polypropylene, 22 gauge, blue, ID 0.016″, 50/box

Application

The tapered design provides smooth flow of viscous fluids and resists clogging when dispensing particle-filled materials.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service