Merck
  • A Tyrosine Switch on NEDD4-2 E3 Ligase Transmits GPCR Inflammatory Signaling.
A Tyrosine Switch on NEDD4-2 E3 Ligase Transmits GPCR Inflammatory Signaling.
Cell reports (2018-09-21)
Neil J Grimsey, Rachan Narala, Cara C Rada, Sohum Mehta, Bryan S Stephens, Irina Kufareva, John Lapek, David J Gonzalez, Tracy M Handel, Jin Zhang, JoAnn Trejo
ABSTRACT
Ubiquitination is essential for protein degradation and signaling and pivotal to many physiological processes. Ubiquitination of a subset of G-protein-coupled receptors (GPCRs) by the E3 ligase NEDD4-2 is required for p38 activation, but how GPCRs activate NEDD4-2 to promote ubiquitin-mediated signaling is not known. Here, we report that the GPCR protease-activated receptor-1 (PAR1) stimulates c-Src-mediated tyrosine phosphorylation and activation of NEDD4-2 to promote p38 signaling and endothelial barrier disruption. Using mass spectrometry, we identified a unique phosphorylated tyrosine (Y)-485 within the 2,3-linker peptide between WW domain 2 and 3 of NEDD4-2 in agonist-stimulated cells. Mutation of NEDD4-2 Y485 impaired E3 ligase activity and failed to rescue PAR1-stimulated p38 activation and endothelial barrier permeability. The purinergic P2Y1 receptor also required c-Src and NEDD4-2 tyrosine phosphorylation for p38 activation. These studies reveal a novel role for c-Src in GPCR-induced NEDD4-2 activation, which is critical for driving ubiquitin-mediated p38 inflammatory signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human NEDD4L
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC₅₀ = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Millipore
ANTI-FLAG® High Sensitivity, M2 coated 96-well plates, 96-well, clear, polystyrene, flat bottom plate
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, 4G10® Platinum, clone 4G10, Upstate®, from mouse