Z764078
MicroTissues® 3D Petri Dish® micro-mold rods
fits 24 well plates, 3 x 8 array
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.14
Recommended Products
material
spherical
description
makes rod-shaped microtissues
sterility
sterile; autoclaved
feature
lid: no
3 x 8 array
packaging
pack of 6 ea
manufacturer/tradename
MicroTissues Inc. 24-24TR
volume
200 μL
Related Categories
General description
Six autoclavable precision micro-molds to cast 3D Petri Dish for forming rod shaped microtissues. 3D Petri Dish for use in 24-well plate. Each micro-mold forms 24 trough shaped recesses in a 3 x 8 array.
- Nominal dimensions of each 3D culture trough: L 2200 μm x D 800 μm x W 400 μm
- Micro-molds also form single chamber for cell seeding
When the gelled agarose is removed from the micro-mold, it is transferred to a standard 12 well or 24 well tissue culture dish and equilibrated with cell culture medium.
Since the agarose is transparent, the spheroids or microtissues that form at the bottom of each agarose micro-well can be easily viewed using a standard inverted microscope using phase contrast, bright field or fluorescence microscopy.
Since the agarose is transparent, the spheroids or microtissues that form at the bottom of each agarose micro-well can be easily viewed using a standard inverted microscope using phase contrast, bright field or fluorescence microscopy.
- The micro-molds are reusable up to 12 times and can be sterilized via a standard steam autoclave (30 min, dry cycle)
- The micro-molds should be stored in a covered container to maintain sterility and to avoid collecting dust or fibres on their small features
- Any microscope with sufficient magnification and resolution functions for cell based views or images will be suitable for use with the 3D Petri Dish products
Legal Information
3D Petri Dish is a registered trademark of MicroTissues Inc.
MicroTissues is a registered trademark of MicroTissues Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)