Skip to Content
Merck
All Photos(1)

Key Documents

SML3644

Sigma-Aldrich

JR-AB2-011

≥98% (HPLC)

Synonym(s):

3-(3,4-Dichlorophenyl)-1-(4-fluorophenyl)-1-(5-methyl-4,5-dihydrothiazol-2-yl)urea, N-(3,4-Dichlorophenyl)-2-[(4-fluorophenyl)imino]-5-methyl-3-thiazolidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14Cl2FN3OS
CAS Number:
Molecular Weight:
398.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

JR-AB2-011 is a selective mTORC2 inhibitor that blocks the interaction of Rictor and mTOR (IC50 = 0.31 µM) without affecing mTORC1 activity or assembly. JR-AB2-011 exhibits anti-glioblastoma (GBM) activity in vitro (proliferation IC50 = 0.31 µM post 72h treatment, 36.3% apoptosis post 48h 1 µM treatment in LN229 cultures) and in vivo (4 or 20 mg/kg/d i.p. to LN229 xenografted mice).
Selective mTORC2 inhibitor with anti-GBM efficacy in vitro and in vivo by blocking Rictor and mTOR interaction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service