Skip to Content
Merck
All Photos(2)

Key Documents

PLA0030

Sigma-Aldrich

Rabbit anti-TopBP1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

DNA topoisomerase II-beta-binding protein 1, DNA topoisomerase II-binding protein 1, KIAA0259, TOP2BP1, TopBP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
species reactivity:
mouse, human
application:
IP
WB
technique(s):
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000-1:10,000
citations:
2

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000-1:10,000

accession no.

NP_008958.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... TopBP1(11073)

Immunogen

The epitope recognized by PLA0030 maps to a region between residue 1400 and the C-terminus (residue 1435) of human Topoisomerase (DNA) II Binding Protein 1 using the numbering given in entry NP_008958.1 (GeneID 11073).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Topoisomerases regulate topological states of DNA. DNA topoisomerase II-binding protein 1 (TopBP1) is a protein that interacts with the C-terminal region of topoisomerase II beta (Topo II beta). TopBP1 function has been found to be important to various aspects of cellular growth control such as DNA replication, the DNA damage checkpoint, and cell survival. TopBP1 interacts with several proteins through its BRCA1 C-terminal (BRCT) motifs. TopBP1 interacts and inhibits E2F1 transcriptional activity through an Rb-independent mechanism that involves recruitment of the SWI/SNF chromatin-remodeling complex proteins Brg1 and Brm.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuying Liu et al.
Nature communications, 8, 15207-15207 (2017-05-11)
Interactions with the immune system may lead tumorigenic cells into dormancy. However, the underlying molecular mechanism is poorly understood. Using a 3D fibrin gel model, we show that IFN-γ induces tumour-repopulating cells (TRCs) to enter dormancy through an indolamine 2,3-dioxygenase
Yuying Liu et al.
Cancer cell, 33(3), 480-494 (2018-03-14)
Despite the clinical successes fostered by immune checkpoint inhibitors, mechanisms underlying PD-1 upregulation in tumor-infiltrating T cells remain an enigma. Here, we show that tumor-repopulating cells (TRCs) drive PD-1 upregulation in CD8+ T cells through a transcellular kynurenine (Kyn)-aryl hydrocarbon receptor (AhR)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service