Key Documents
CLS431845
Corning® centrifuge bottle
with standard screw cap, capacity 500 mL, polypropylene
Synonym(s):
centrifugation bottles, digestion bottles, shaking bottles
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Pricing and availability is not currently available.
material
flat bottom tube
polypropylene
round bottle
translucent bottle
wide-mouth bottle
sterility
non-sterile
feature
autoclavable
cap
packaging
pkg of 24 ea
manufacturer/tradename
Corning 431845
parameter
13700 × g max. RCF
capacity
500 mL
Features and Benefits
  • Corning® centrifuge bottles have a specially designed cap for improved grip
  • Includes clear graduations and textured marking spot.
  • Bottles are designed with extra-thick walls for high durability
  • Wide mouth for easier liquid filling
  • Compatible with most centrifuges and do not require special buckets
  • Unique design aid researchers to reduce the number of centrifugation cycles and enhance productivity
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)