CLS4133
Corning® DeckWorks™ pipet tips reload starter kits
1 - 300μL pipet tips, graduated, reload starter kit, natural, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 960 tips/pack, 1 packcase and 10 empty racks/case
Synonym(s):
pipette tip
All Photos(1)
About This Item
Recommended Products
material
natural tip
polypropylene
description
Starter Kit
sterility
non-sterile
feature
barrier: no
disposable
graduated
Tip format: 96 tips/rack
Tip shape: round
packaging
pack of 960
manufacturer/tradename
Corning 4133
volume range
1-300 μL
General description
Corning® DeckWorks™ Reload Starter Kits
DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks. 10 empty racks accompany 10 uL, 200 uL and 300 uL volume tips and 8 empty racks accompany 1000 uL volume tips.
This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic.
The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.
Medical grade resins and production excellence allow for full certification as:
DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks. 10 empty racks accompany 10 uL, 200 uL and 300 uL volume tips and 8 empty racks accompany 1000 uL volume tips.
This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic.
The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.
Medical grade resins and production excellence allow for full certification as:
- Nonpyrogenic
- Human DNA-free
- RNase-/DNase-free
Legal Information
Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.