Skip to Content
Merck
All Photos(2)

Key Documents

MAB19562-AF647

Sigma-Aldrich

Anti-Laminin-5 Antibody (γ2 chain), clone D4B5, Alexa Fluor 647 Conjugate

clone D4B5, from mouse, ALEXA FLUOR 647

Synonym(s):

Laminin subunit gamma-2, Cell-scattering factor 140 kDa subunit, CSF 140 kDa subunit, Epiligrin subunit gamma, Kalinin subunit gamma, Kalinin/nicein/epiligrin 100 kDa subunit, Ladsin 140 kDa subunit, Laminin B2t chain, Laminin-5 subunit gamma, Large adhe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified antibody

antibody product type

primary antibodies

clone

D4B5, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LAMC2(3918)

General description

Laminin subunit gamma-2 (UniProt Q13753; also known as CSF 140 kDa subunit, Cell-scattering factor 140 kDa subunit, Epiligrin subunit gamma, Kalinin subunit gamma, Ladsin 140 kDa subunit, Laminin B2t chain, Lamin-5 subunit gamma, Large adhesive scatter factor 140 kDa subunit, Nicein subunit gamma) is encoded by the LAMC2 (also known as B2T, LAMB2T, LAMNB2) gene (Gene ID 3918) in human. Laminins are basement membrane, extracellular matrix glycoproteins composed of alpha, beta and gamma heterotrimeric chains. Different combinations of five alpha chains, three beta chains, and two gamma chains form at least 12 laminin isoforms. Laminin-5 (LN5), which consists of laminin alpha3, beta3 and gamma2 chains, is a highly conserved component of anchoring filaments underlying the hemidesmosome structure of epidermal keratinocytes. Mutations or deletion in the LN5 genes (LAMA3, LAMB3, and LAMC2) is associated with epidermolysis bullosa, a lethal skin blistering disease. LN5 may stabilize the epidermal/dermal junction of skin through interaction with integrins alpha3/beta1, alpha6/beta4 and type VII collagen and has a critical role in the anchorage and locomotion of a wide variety of cell types, including epithelial cells, fibroblasts, neurons and leukocytes. Comparing to fibronectin, collagen, and vitronectin, cells of epithelial origin will adhere to Laminin-5 faster and will spread to a larger extent. Laminin-5 attracted attention when it was detected at the leading edge of invasive cancer tissue in several types of human carcinomas (Niki, T., et al. (2002). Am. J. Pathol. 160(3):112911-41; Lohi, J. (2001). Int. J. Cancer. 94(6):763-767).

Specificity

This antibody recognizes the 150 (precursor form) and 105 kDa (mature form) chain proteins of Laminin-5. No reactivity is expected with other laminin types, as the gamma2 chain of laminin is unique to laminin-5.

Immunogen

Amino acid residues 382-608 of human laminin-5 gamma2 chain (III domain) expressed as a GST fusion protein.
Epitope: EGF-like 4 (second part)

Application

Anti-Laminin-5 Antibody (γ2 chain), clone D4B5, Alexa Fluor 647 Conjugate is an antibody against Laminin-5 for use in Immunocytochemistry.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Laminin-5 in NIH/3T3 cells.
The unconjugated antibody (Cat. No. MAB19562) is shown to be suitable also for Western blotting, ELISA, and immunohistochemistry applications.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Laminin-5 in A431 cells.

Target description

~150 kDa calculated

Physical form

Protein A purified
Purified mouse antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priscilla Y Hwang et al.
Developmental cell, 58(1), 34-50 (2023-01-11)
Carcinoma dissemination can occur when heterogeneous tumor and tumor-stromal cell clusters migrate together via collective migration. Cells at the front lead and direct collective migration, yet how these leader cells form and direct migration are not fully appreciated. From live

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service