Skip to Content
Merck
All Photos(1)

Key Documents

AB5636P

Sigma-Aldrich

Anti-Cannabinoid Receptor 1 Antibody, NT

Chemicon®, from rabbit

Synonym(s):

CB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CNR1(1268)
mouse ... Cnr1(12801)

Specificity

Recognizes human Cannabinoid Receptor 1 (CB1). The immunogen shows no significant sequence homology with CB2 or other receptors.

Immunogen

A 14 amino acid N-terminus peptide sequence (aa 1-14) (Gerard et al. 1991; Gerard et al.

1990; Shire et al. 1995; Matsuda et al. 1990; Ho et al. 1996; Chakraborty et al. 1995).
Epitope: N-terminus

Application

Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Neurotransmitters & Receptors
This Anti-Cannabinoid Receptor 1 Antibody, N-terminus is validated for use in ELISA, WB for the detection of Cannabinoid Receptor 1.
Western blot: 1-10 μg/mL using ECL. Expected sizes are ~60kDa and 80-84kDa under reducing conditions.

ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide (Cat. No. AG357) per well.

Optimal working dilutions must be determined by the end user.

Target description

~60kDa and 80-84kDa

Physical form

Affinity Purified immunoglobulin. Liquid in 100mM Tris, pH 7.5, containing 0.2% BSA and 0.05% sodium azide.
ImmunoAffinity Purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angel Arévalo-Martín et al.
The European journal of neuroscience, 26(6), 1548-1559 (2007-09-21)
The subventricular zone (SVZ) is a source of post-natal glial precursors that can migrate to the overlying white matter, where they may differentiate into oligodendrocytes. We showed that, in the post-natal SVZ ependymocytes, radial glia and astrocyte-like cells express cannabinoid
Cannabinoid control of brain bioenergetics: Exploring the subcellular localization of the CB1 receptor.
Hebert-Chatelain, E; Reguero, L; Puente, N; Lutz, B; Chaouloff, F; Rossignol, R; Piazza et al.
Molecular Metabolism null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service