Skip to Content
Merck
All Photos(1)

Key Documents

1.05596

Supelco

Sodium hydroxide solution

about 36%, for analysis EMSURE®

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Sodium hydroxide solution about 36%, for analysis EMSURE®

grade

reagent

Quality Level

product line

EMSURE®

form

liquid

concentration

36.0-36.5% NaOH

impurities

≤0.00035% Fe (Iron)
≤0.005% Chloride (Cl)
≤1.0% Carbonate (as Na2CO3)

pH

14 (20 °C in H2O)

density

1.39 g/cm3 at 20 °C

storage temp.

15-25°C

General description

Sodium hydroxide solutionabout 36% (EMSURE® grade product) is used for regulated analyses and highlydemanding lab applications. It also offers the highest quality and unmatchedscope of specifications to give complete control of test conditions andeliminate uncertainties. EMSURE® grade products comply with all internationalregulations and can be used in various applications.

Application

Sodium hydroxidesolution about 36% can be used as an additive in the selective silica removalstrategy to enable raw material extraction from geothermal fluids.

Features and Benefits

  • Most extensive specifications worldwide
  • Tested for up to 70 parameters
  • Extraordinary purity
  • Very low limiting values
  • Greater accuracy and control of analyses
  • Optimized for highly critical and demanding analyses
  • Ideal for method development
  • No interference or contamination due to unknown impurities

Analysis Note

Assay (acidimetric,NaOH)36.0 - 36.5 %Carbonate (as Na₂CO₃)≤ 1.0 %Chloride (Cl)≤ 0.005 %Fe (Iron)≤ 0.00035 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service