Skip to Content
Merck
All Photos(1)

Key Documents

900120O

Avanti

PGB1

Avanti Research - A Croda Brand 900120O

Synonym(s):

Prostaglandin B1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O4
CAS Number:
Molecular Weight:
336.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mg (900120O-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900120O

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C1C(CCCCCCC(O)=O)=C(/C=C/[C@H](CCCCC)O)CC1

InChI

1S/C20H32O4/c1-2-3-6-9-17(21)14-12-16-13-15-19(22)18(16)10-7-4-5-8-11-20(23)24/h12,14,17,21H,2-11,13,15H2,1H3,(H,23,24)/b14-12+/t17-/m0/s1

InChI key

YBHMPNRDOVPQIN-VSOYFRJCSA-N

General description

Prostaglandins are hormones present in many tissues and organs.Prostaglandin B1 (PGB1) is a ketone molecule with a five carbon ring structure. It also contains two side chains linked to its unsaturated region.

Biochem/physiol Actions

Prostaglandin B1 (PGB1) is a metabolic product of PGE1 dehydration. It is known to specifically bind to peroxisome proliferator-activated receptor-γ. By this, PGB1 mediates fat accumulation and metabolism. Oligomerization of PGB1 potentiates its anti-oxidative property.

Packaging

5 mL Clear Glass Sealed Ampule (900120O-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General Approach to Prostanes B1 by Intermolecular Pauson-Khand Reaction: Syntheses of Methyl Esters of Prostaglandin B1 and Phytoprostanes 16-B1-PhytoP and 9-L1-PhytoP
Vazquez RA, et al.
European Journal of Organic Chemistry, 2013(9), 1716-1725 (2013)
Synthesis of Prostaglandin and Phytoprostane B1 Via Regioselective Intermolecular Pauson- Khand Reactions
Vazquez RA, et al.
Organic Letters, 11(14), 3104-3107 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service