Z268623
Scienceware® Safe-Lab™ PTFE stopper
joint: ST/NS 29/42, hollow
Synonym(s):
lab stopper, lab stoppers, stoppers
All Photos(1)
About This Item
UNSPSC Code:
41122500
NACRES:
NB.35
Joint
joint: ST/NS29/42
packaging
pkg of 2 ea
manufacturer/tradename
Bel-Art F20197-1530
General description
PTFE stoppers with PP extracting nut for easy removal from flask. For use with boiling and distilling flasks with ground joints.
Legal Information
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)