Documents
Z115746
Sigma-Aldrich
Aldrich® vacuum-transfer adapter
joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
joint: ST/NS24/40
stopcock size
4 mm
General description
PTFE stopcocks. For transfer of hazardous and sensitive reagents.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC