GF96129760
Aluminum-silicon alloy, Al99Si1
foil, 100 x 100mm, 6.0mm thickness, as rolled, 99.998%
Synonym(s):
AL050680
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352311
NACRES:
NA.23
Assay
99.998% (high purity)
form
foil (as rolled)
manufacturer/tradename
Goodfellow 961-297-60
size × thickness
100 mm × 100 mm × 6.0 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service