Skip to Content
Merck
All Photos(1)

Key Documents

GF60413217

Dural® - aluminum-copper-magnesium alloy, Al95Cu4Mg1

100mm tube, 0.53mm outside diameter, 0.051mm wall thickness, 0.428mm inside diameter, as drawn

Synonym(s):

AL047105

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 604-132-17

L × wall thickness

100 mm × 0.051 mm

O.D. × I.D.

0.53 mm × 0.428 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Dural is a registered trademark of Lonza-Werke GmbH
Dural is a registered trademark of Lonza-Werke GmbH

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service