Skip to Content
Merck
All Photos(1)

Key Documents

GF22306676

Havar® - high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn

foil, 50 x 50mm, 0.075mm thickness, as rolled, light tight (lt)

Synonym(s):

CO010300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

Pricing and availability is not currently available.

description

light tight (lt)

form

foil (as rolled)

manufacturer/tradename

Goodfellow 223-066-76

size × thickness

50 mm × 50 mm × 0.075 mm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF20360326GF07336807GF06410292
manufacturer/tradename

Goodfellow 223-066-76

manufacturer/tradename

Goodfellow 203-603-26

manufacturer/tradename

Goodfellow 073-368-07

manufacturer/tradename

Goodfellow 064-102-92

description

light tight (lt)

description

light tight (lt)

description

-

description

-

size × thickness

50 mm × 50 mm × 0.075 mm

size × thickness

25 mm × 25 mm × 0.035 mm

size × thickness

50 mm × 50 mm × 0.2 mm

size × thickness

50 mm × 50 mm × 0.5 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Havar is a registered trademark of Precision Metals, Inc. AMETEK, or its subsidiaries or affiliates.
Havar is a registered trademark of Precision Metals, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service