BR143265
BRAND® special laboratory tubing
I.D. × O.D. × W 5 mm × 8 mm × 1.5 mm, L 20 m
Synonym(s):
Special laboratory tubing, Tubing
All Photos(1)
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
Recommended Products
material
PVC
manufacturer/tradename
BRAND 143265
I.D. × O.D. × W
5 mm × 8 mm × 1.5 mm
L
20 m
Related Categories
General description
Particularly high-grade material, manufactured to a first-class standard. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies. Autoclavable (121 °C).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)