Skip to Content
Merck
All Photos(2)

Key Documents

571571

Sigma-Aldrich

Potassium 3-thiophenetrifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3BF3KS
CAS Number:
Molecular Weight:
190.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

SMILES string

[K+].F[B-](F)(F)c1ccsc1

InChI

1S/C4H3BF3S.K/c6-5(7,8)4-1-2-9-3-4;/h1-3H;/q-1;+1

InChI key

ULOOPHJJEPUNNH-UHFFFAOYSA-N

General description

Potassium 3-thiophenetrifluoroborate is a nucleophilic boronated coupling reagent used to construct a C-C bond by reacting with aryl halides in the presence of a catalyst or under thermal conditions.

Application

Potassium 3-thiophenetrifluoroborate can be used as a reactant to prepare:
  • Methyl 3-amino-6-(thien-3-yl)thieno[3,2-b]pyridine-2-carboxylate by reacting with 6-bromothieno[3,2-b]pyridine.
  • Thiophene derivatives via Suzuki-Miyaura cross-coupling reaction with aryl halides using palladium complex as a catalyst.
  • 2-(Thiophen-3-yl)-3,1,2-benzoxazaborininone and 2-(thiophen-3-yl)-1,3,2-benzoxazaborininone by treating with 2-aminobenzoic acid and 2-hydroxybenzamide, respectively.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzuki-Miyaura Cross-Coupling Reaction Catalyzed by Palladium Complexes of Hydroxynaphthalene-2-Oxazolines
Barbeiro CS, et at.
ChemistrySelect, 2(26), 8173-8177 (2017)
Efficient synthesis of 6-(hetero) arylthieno [3, 2-b] pyridines by Suzuki-Miyaura coupling. Evaluation of growth inhibition on human tumor cell lines, SARs and effects on the cell cycle
Queiroz M-J, et al.
European Journal of Medicinal Chemistry, 45(12), 5628-5634 (2010)
Azaborininones: Synthesis and Structural Analysis of a Carbonyl-Containing Class of Azaborines
Davies GHM, et al.
The Journal of Organic Chemistry, 82(10), 5380-5390 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service