Skip to Content
Merck
All Photos(1)

Key Documents

336467

Sigma-Aldrich

Benz[cd]indol-2(1H)-one

technical grade

Synonym(s):

NSC 25094

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7NO
CAS Number:
Molecular Weight:
169.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

solid

mp

173-178 °C (lit.)

solubility

methanol: soluble 25 mg/mL, clear to slightly hazy, yellow to brown

SMILES string

O=C1Nc2cccc3cccc1c23

InChI

1S/C11H7NO/c13-11-8-5-1-3-7-4-2-6-9(12-11)10(7)8/h1-6H,(H,12,13)

InChI key

GPYLCFQEKPUWLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benz[cd]indol-2(1H)-one is also referred as naphtholactam.

Application

  • Reactant in photo-Fries rearrangement
  • Reactant in preparation of potential antitumor agents
  • Reactant in synthesis of inhibitors of thymidylate synthase
Benz[cd]indol-2(1H)-one was used in the synthesis of 2-oxo-1,2-dihydrobenzo[cd]indole-6-sulfonamide analog, novel Mycobacterium protein tyrosine phosphatase B (mPTPB) inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Saeed I Khan et al.
Acta crystallographica. Section C, Crystal structure communications, 68(Pt 1), o1-o6 (2012-01-10)
Weakly diffracting crystals of benz[cd]indol-2(1H)-one (naphtholactam), C(11)H(7)NO, were unsuitable for data collection by early photographic methods. However, a diffractometer data set collected at room temperature in 1989 was solved and refined. The peak scans were broad, and the results indicated
Lan Chen et al.
ACS medicinal chemistry letters, 1(7), 355-359 (2010-12-01)
Mycobacterium protein tyrosine phosphatase B (mPTPB) is an essential virulence factor required for Mycobacterium tuberculosis (Mtb) survival in host macrophages. Consequently, mPTPB represents an exciting new target with a completely novel mechanism of action. We screened a library of 7,500

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service