Principaux documents
Z126063
Ace McLeod vacuum gauge
pressure 0-15 mmHg
About This Item
Code UNSPSC :
41112406
Nomenclature NACRES :
NB.79
Fabricant/nom de marque
Ace Glass 872809
Paramètres
0-15 mmHg pressure
Description générale
With interchangeable measuring tube. Mercury not included.