Principaux documents
BR701365
BRAND® sealing film for microplates
self-adhesive, gas permeable, suitable for (for cell and tissue culture)
Synonyme(s) :
Accessories, PCR, Plates, Sealing films
About This Item
Code UNSPSC :
41122410
Nomenclature NACRES :
NB.22
Matériaux
Rayon
Rayon film
Stérilité
sterile
Caractéristiques
gas permeable
self-adhesive
Conditionnement
pack of 50 ea
Fabricant/nom de marque
BRAND 701365
Adéquation
suitable for (for cell and tissue culture)
Description générale
When working with cell and tissue cultures, ensuring a secure seal is only the first step in protecting against contamination and facilitating safe transport. Equally critical is the ability to allow for proper gas exchange. Insufficient oxygen levels or excessive carbon dioxide can not only compromise the accuracy of experimental results but may also jeopardize cell viability. Consequently, films designed for cell and tissue cultures must meet stringent requirements to support both secure sealing and optimal gas exchange, ensuring a conducive environment for cellular growth and experimentation.
Caractéristiques et avantages
• Air permeable for optimum oxygen supply• Secure seal against contamination• Pierceability for easy sample extraction
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG