MilliporeSigma
  • A Luminescent Nitrogen-Containing Polycyclic Aromatic Hydrocarbon Synthesized by Photocyclodehydrogenation with Unprecedented Regioselectivity.
A Luminescent Nitrogen-Containing Polycyclic Aromatic Hydrocarbon Synthesized by Photocyclodehydrogenation with Unprecedented Regioselectivity.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-10-23)
Xinggui Gu, Hong Wang, Jesse Roose, Zikai He, Yue Zhou, Yongli Yan, Yuanjing Cai, Heping Shi, Yilin Zhang, Herman H Y Sung, Jacky W Y Lam, Qian Miao, Yongsheng Zhao, Kam Sing Wong, Ian D Williams, Ben Zhong Tang
ABSTRACT
We present a nitrogen-containing polycyclic aromatic hydrocarbon (N-PAH), namely 12-methoxy-9-(4-methoxyphenyl)-5,8-diphenyl-4-(pyridin-4-yl)pyreno[1,10,9-h,i,j]isoquinoline (c-TPE-ON), which exhibits high quantum-yield emission both in solution (blue) and in the solid state (yellow). This molecule was unexpectedly obtained by a three-fold, highly regioselective photocyclodehydrogenation of a tetraphenylethylene-derived AIEgen. Based on manifold approaches involving UV/Vis, photoluminescence, and NMR spectroscopy as well as HRMS, we propose a reasonable mechanism for the formation of the disk-like N-PAH that is supported by density functional theory calculations. In contrast to most PAHs that are commonly used, our system does not suffer from entire fluorescence quenching in the solid state due to the peripheral aromatic rings preventing π-π stacking interactions, as evidenced by single-crystal X-ray analysis. Moreover, its rod-like microcrystals exhibit excellent optical waveguide properties. Hence, c-TPE-ON comprises a N-PAH with unprecedented luminescent properties and as such is a promising candidate for fabricating organic optoelectronic devices. Our design and synthetic strategy might lead to a more general approach to the preparation of solution- and solid-state luminescent PAHs.
Sigma-Aldrich
4,4′-Dimethoxybenzophenone, 97%