Merck
Serum biomarkers predictive of pre-eclampsia.
Biomarkers in medicine (2015-06-17)
Swati Anand, Tanielle Mei Bench Alvarez, W Evan Johnson, M Sean Esplin, Karen Merrell, T Flint Porter, Steven W Graves
ABSTRACT
We sought serum biomarkers predictive of pre-eclampsia (PE). Sera obtained at 12-14 weeks of pregnancy from 24 cases who later developed PE and 24 controls with uncomplicated pregnancies were processed and analyzed using a serum proteomic approach. Many statistically significant serum PE biomarker candidates (n > 60) were found comparing cases and controls. In addition, logistic regression analysis modeled biomarker data resulted in 14 different multimarker combinations having high detection sensitivity and specificity (AUC >0.9). Developed panels of serum biomarkers appeared effective in identifying pregnant women at 12-14 weeks gestation at risk of PE later in their pregnancy.
Sigma-Aldrich
1-Oleoyl-sn-glycero-3-phosphocholine, synthetic, ≥99%