Merck
1-[3,5-Bis(trifluoro-meth-yl)phen-yl]-3-(2-pyrid-yl)thio-urea.
Acta crystallographica. Section E, Structure reports online (2008-01-01)
Huadong Yue, Yifeng Wang, Aibao Xia, Shuping Luo, Danqian Xu
ABSTRACT
The title compound, C(14)H(9)F(6)N(3)S, exhibits a nearly planar conformation in the solid state, with a dihedral angle between the planes of the benzene and pyridine rings of 14.86 (3)°. The pyridine N atom allows for the formation of a six-membered N-H⋯N(py) hydrogen-bonded ring, thus forcing the two amide H atoms of the thio-urea group to point in opposite directions. The second N-H group forms an inter-molecular N-H⋯S hydrogen bond with the S atom of an adjacent mol-ecule. The F atoms of the two trifluoro-methyl groups display rotational disorder around the C-CF(3) axis, with an occupancy ratio of 0.54 (1):0.46 (1).
Sigma-Aldrich
3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%