Merck
  • Unusual formation of single-crystal manganese sulfide microboxes co-mediated by the cubic crystal structure and shape.
Unusual formation of single-crystal manganese sulfide microboxes co-mediated by the cubic crystal structure and shape.
Angewandte Chemie (International ed. in English) (2012-06-12)
Lei Zhang, Liang Zhou, Hao Bin Wu, Rong Xu, Xiong Wen David Lou
ABSTRACT
Kept cubic: MnS microboxes, which act as an anode material for lithium ion batteries, are synthesized by a simple H(2)S gas sulfidation approach (TEM images show porous and hollow microcubes and a microbox). The formation of the single crystals is aided by the intrinsic cubic crystal structure and the nearly cubic shape of the MnCO(3) precursor.
Sigma-Aldrich
Manganese(II) sulfide, ~100 mesh particle size
Pricing and availability is not currently available.