Merck
  • Generalized cytochrome P450-mediated oxidation and oxygenation reactions in aromatic substrates with activated N-H, O-H, C-H, or S-H substituents.
Generalized cytochrome P450-mediated oxidation and oxygenation reactions in aromatic substrates with activated N-H, O-H, C-H, or S-H substituents.
Xenobiotica; the fate of foreign compounds in biological systems (1993-06-01)
L Koymans, G M Donné-Op den Kelder, J M te Koppele, N P Vermeulen
ABSTRACT
1. The general mechanism of metabolic oxidation of substrates by cytochromes P450 (P450s) appears to consist of sequential one-electron oxidation steps rather than of a single concerted transfer of activated oxygen species from P450 to substrates. 2. In case of the acetanilides paracetamol (PAR), phenacetin (PHEN), and 4-chloro-acetanilide (4-CLAA), the first one-electron oxidation step consists of a hydrogen abstraction from the acetylamino nitrogen and/or from the other side-chain substituent on the aromatic ring. The substrate radicals thus formed delocalize their spin and the respective reactive centres of the substrate radical recombine with a P450 iron-bound hydroxyl radical to either yield oxygenated metabolites, or undergo a second hydrogen abstraction forming dehydrogenated products. By this mechanism, the formation of all known oxidative metabolites of PAR, PHEN, and 4-ClAA can be explained. Furthermore, this mechanism is consistent with all available experimental data on [18O]PAR/PHEN, [2H]PAR, and [14C]PHEN. 3. The oxidative metabolic reactions proposed for the acetanilides PAR, PHEN, and 4-ClAA are used to generalize P450-mediated oxidations of these and other acetanilides, such as analogues of PAR and 2-N-acetyl-aminofluorene. 4. A further generalization of the hydrogen abstraction, spin delocalization, radical recombination concept is derived for other aromatic substrates with abstractable hydrogen atoms, notably those with activated N-H, O-H, C-H, or S-H bonds directly attached to the aromatic nucleus.
Sigma-Aldrich
4′-Chloroacetanilide, 97%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetaminophen Related Compound J, Pharmaceutical Secondary Standard; Certified Reference Material