Merck
  • Whitening Activity of Constituents Isolated from the Trichosanthes Pulp.
Whitening Activity of Constituents Isolated from the Trichosanthes Pulp.
Evidence-based complementary and alternative medicine : eCAM (2020-08-11)
Rongchao Zhang, Xiuqin Hu, Bo Zhang, Zhen Wang, Chunxiang Hao, Jie Xin, Qingmei Guo
ABSTRACT
Whitening cosmetics market has a bright future, and pure natural whitening products of traditional Chinese medicine have always been a research hotspot. In this research, the whitening active ingredient of Chinese medicine Trichosanthes pulp was isolated and purified for the first time, and its whitening mechanism was clarified. Chromatographic methods such as silica gel, ODS, and HPLC were used to isolate and purify them. B16 cells were used to measure the antioxidant activity, tyrosinase activity, and melanin removal activity. A total of 20 compounds were isolated, including p-hydroxybenzaldehyde (1), salicylic acid (2), vanillic acid (3), isovanillic acid (4), protocatechuate (5), trans-cinnamic acid (6), 4-coumaric acid (7), trans-ferulic acid (8), drechslerol-B (9), cyclotucanol 3-palmitate (10), 5-acetoxymethyl-2-furaldehyde (11), 5-hydroxymethylfurfural (12), diosmetin (13), apigenin (14), chrysoeriol (15), luteolin (16), 4'-hydroxyscutellarin (17), quercetin (18), 3',5-dihydroxy-7-(β-D-glucopyranosyloxy)-4'-methoxyflavone (19), and cofloxacin-7-O-β-D-glucoside (20). Among them, compounds 9, 10, 11, 12, 13, 14, 15, 16, 17, 18, 19, and 20 have good antioxidant repairing effects; compounds 3, 4, 5, 6, and 7 have high black inhibition; compounds 1, 2, 3, 4, 5, 6, 7, 8, 11, 12, 13, 14, 15, 16, 17, 18, 19, and 20 have obvious tyrosine acidase inhibitory activity. The results laid foundation for the further development and utilization of Trichosanthes pulp resources and also provide a basis for the development of natural whitening cosmetics.
Sigma-Aldrich
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid