Merck
  • Functional analysis of Tcl1 using Tcl1-deficient mouse embryonic stem cells.
Functional analysis of Tcl1 using Tcl1-deficient mouse embryonic stem cells.
PloS one (2013-08-14)
Tatsushi Miyazaki, Satsuki Miyazaki, Masafumi Ashida, Tomofumi Tanaka, Fumi Tashiro, Jun-ichi Miyazaki
ABSTRACT
Tcl1 is highly expressed in embryonic stem (ES) cells, but its expression rapidly decreases following differentiation. To assess Tcl1's roles in ES cells, we generated Tcl1-deficient and -overexpressing mouse ES cell lines. We found that Tcl1 was neither essential nor sufficient for maintaining the undifferentiated state. Tcl1 is reported to activate Akt and to enhance cell proliferation. We found that Tcl1 expression levels correlated positively with the proliferation rate and negatively with the apoptosis of ES cells, but did not affect Akt phosphorylation. On the other hand, the phosphorylation level of β-catenin decreased in response to Tcl1 overexpression. We measured the β-catenin activity using the TOPflash reporter assay, and found that wild-type ES cells had low activity, which Tcl1 overexpression enhanced 1.8-fold. When the canonical Wnt signaling is activated by β-catenin stabilization, it reportedly helps maintain ES cells in the undifferentiated state. We then performed DNA microarray analyses between the Tcl1-deficient and -expressing ES cells. The results revealed that Tcl1 expression downregulated a distinct group of genes, including Ndp52, whose expression is very high in blastocysts but reduced in the primitive ectoderm. Based on these results, we discuss the possible roles of Tcl1 in ES cells.
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glasgow Minimum Essential Medium, With L-glutamine, without tryptose phosphate broth and sodium bicarbonate, powder, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TCF Reporter Plasmid Kit, Set of transfection grade T cell factor (TCF) reporter plasmids for use in TOPFlash and FOPFlash wnt/b-catenin activity assays.