Skip to Content
Merck
All Photos(1)

Key Documents

PHL80712

Hydroquinone

phyproof® Reference Substance

Synonym(s):

1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
605970
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

vapor density

3.81 (vs air)

vapor pressure

1 mmHg ( 132 °C)

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

form

crystals

autoignition temp.

930 °F

manufacturer/tradename

PhytoLab

bp

285 °C (lit.)

mp

172-175 °C (lit.)

SMILES string

Oc1ccc(O)cc1

InChI

1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H

InChI key

QIGBRXMKCJKVMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

329.0 °F - closed cup

Flash Point(C)

165 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service