Skip to Content
Merck
All Photos(1)

Key Documents

58970C30

Supelco

SUPELCOSIL LC-18-T (3 µm) HPLC Columns

L × I.D. 15 cm × 3 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

SUPELCOSIL LC-18-T HPLC Column, 3 μm particle size, L × I.D. 15 cm × 3 mm

material

stainless steel column

Agency

suitable for USP L1

product line

SUPELCOSIL

feature

endcapped

manufacturer/tradename

SUPELCOSIL

packaging

1 ea of

extent of labeling

12.3% carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 3 mm

surface area

170 m2/g

surface coverage

surface coverage 3.1 μmol/m2

matrix

silica gel, spherical particle platform
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

3 μm

pore size

120 Å

operating pH

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL LC-18-T columns feature an octadecylsilane bonded phase and a special surface treatment for efficient separations of nucleotides. Each batch of packing material is tested to ensure good peak shape for a representative nucleotide, adenosine diphosphate (ADP). Chromatography of other compounds that exhibit metal chelating properties also can be improved by using this phase.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khaled S Abdelkawy et al.
Biomedical chromatography : BMC, 31(3) (2016-08-25)
A simple, accurate, and reproducible high-performance liquid chromatography (HPLC) method has been developed and validated for the quantification of quercetin (QR) in rat plasma. The method involves a simple protein precipitation procedure to extract both QR and thymoquinone (TQ), the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service