52791-U
Empty polypropylene SPE Tube with PE frits
Synonym(s):
Empty polypropylene SPE Tube with PE frits, 20 μm porosity
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104018
NACRES:
NB.21
Recommended Products
material
polypropylene
description
includes polyethylene frits: 20 μm pore size
packaging
pk of 5000
technique(s)
solid phase extraction (SPE): suitable
volume
3 mL
pore size
20 μm
application(s)
food and beverages
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Empty SPE Tubes allow you to pack your own SPE cartridges
Other Notes
Frits are included separately, pk of 10,000.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service