Z740130
BRAND® TipBox, empty
with blue tip-tray for 1000 μL tips
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.01
Recommended Products
material
colorless polypropylene box, lid, and holder (includes a colored tip-tray)
sterility
non-sterile
feature
autoclavable (121°C, 2 bar)
Holds <1000 μl tips
stackable
packaging
box of 1
manufacturer/tradename
BRAND 732996
General description
TipBox, non-sterile: the 5 mL and 10 mL tips are only available as a racked tip version in these specially corresponding TipBoxes.
Other Notes
BR732996 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG