Z688657
Nunclon® Δ tube with flat side
sterile
Synonym(s):
cell culture tube, surfce certified tube
All Photos(1)
About This Item
UNSPSC Code:
41121706
NACRES:
NB.22
Recommended Products
material
colorless tube
polyethylene cap
polystyrene tube
round bottom
screw top cap
sterility
sterile
feature
neck thread external
packaging
pack of 450 ea
manufacturer/tradename
Nunc 156758
L × O.D.
110 mm × 16 mm
capacity
3 mL
General description
Nunclon® Δ surface certified tubes have a flat side which allows for easy microscopy, is useful for culture of adherent cells and has excellent optical quality.
- Round bottom version with screw or push-on cap
- Coverslip can be used in the tube
- Medium retained in the flat sided tube in horizontal position
Legal Information
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries