Key Documents
Z380326
Jars Nalgene® style 2117
capacity 500 mL
Synonym(s):
laboratory jars, nalgene jars
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
polymethylpentene bottle
polypropylene cap
polypropylene screw closure
round transparent bottle
Joint
Joints threaded neck (120)
description
Autoclavable with closure completely unscrewed
feature
with cap
packaging
pkg of 4
manufacturer/tradename
Nalgene® 2117-0500
bottle diam. × H
112 mm × 88 mm
capacity
500 mL
General description
Transparent, with good chemical resistance.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
  • Certificates of Analysis (COA)
    Need A Sample COA?
    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.