VIV102005
SynVivo SynBBB 3D blood brain barrier model chip, IMN2 radial configuration
About This Item
Recommended Products
packaging
case of 1 ea
manufacturer/tradename
102005
SynVivo 102005
storage temp.
room temp
1 of 4
This Item | 46935 | 21888 | F9551 |
---|---|---|---|
λex 543 nm; λem 575 nm in 0.1 M phosphate pH 7.0 | λex 491 nm; λem 515 nm in 0.1 M Tris pH 9.0 | λex 492 nm; λem 517 nm in 0.1 M Tris pH 8.0 (esterase) | λex 511-519 nm |
powder | solid | solid | powder |
diagnostic assay manufacturing | - | - | diagnostic assay manufacturing |
DMF: 2 mg/mL, clear, acetonitrile: soluble | DMF: soluble, DMSO: soluble | DMF: soluble, DMSO: soluble | DMSO or DMF: soluble |
−20°C | 2-8°C | −20°C | −20°C |
General description
Linkage
application text
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service