Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥75%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2
Application
Calcitonin, a 32 amino acid polypeptide, is involved in the regulation of calcium and phosphorous metabolism often antagonistic to parathyroid hormone (PTH). [Des-Cys1]-Calcitonin, salmon (Des-Cys1-sCT) has the N-terminal cysteine residue removed.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Biochemical and biophysical research communications, 187(1), 8-13 (1992-08-31)
Structural requirements for binding to the bone calcitonin (CT) receptor and for CT bioactivity both in vitro and in vivo were assessed for a series of N-terminally truncated, N alpha-acetylated, fragments of salmon calcitonin (sCT). Sequential deletion of amino acid
Biological & pharmaceutical bulletin, 22(3), 244-252 (1999-04-29)
Various derivatives of human calcitonin have been synthesized and their biological characteristics compared with those of existing calcitonins. The acute effects of these analogues in reducing serum calcium levels and suppressing appetite were assessed in rats. A calcitonin analogue, PO-1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service