HS120070
TrueNorth® Mini Cooler
for tubes, 0.5 ‑ 2 mL, with clear lid, temp. limit 0 °C, size 12 wells, green polycarbonate
Synonym(s):
cooling tube rack, tube chiller
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103025
NACRES:
NB.22
Recommended Products
material
green polycarbonate
feature
for tubes 0.5 ‑ 2 mL
parameter
0 °C temp. limit
L × W × H
145 × 105 × 100 mm
size
12 wells
Looking for similar products? Visit Product Comparison Guide
General description
Protect temperature sensitive samples at the bench or in the freezer. Mini Coolers are ideal for keeping PCR reagents, enzymes, and other biochemical reagents cool while working at the bench. Filled with insulating gel, the mini coolers can also be used for ccontrolled sample freezing and for longer temperature retention samples.
Features and Benefits
- polycarbonate construction with rubber non-slip feet
- gridded and keyed one-position lids for easy sample identification
- 0.5 mL tube adapters supplied as standard with the 0.5-2.0 mL tube size versions
- temperature retention graph supplied with each of the 7 styles
- versions available or 0.5 and 1.5/2.0 mL microcentrifuge tubes and cyrogenic vials and a 96-place version for 0.2 mL tubes and assay plates
Legal Information
TrueNorth is a registered trademark of Heathrow Scientific LLC
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service