Skip to Content
Merck
All Photos(1)

Key Documents

GE10600094

Amersham Protran® Western blotting membranes, nitrocellulose

pore size 0.2 μm, sheet W × L 80 mm × 90 mm, pkg of 25 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

Pricing and availability is not currently available.

packaging

pkg of 25 ea

manufacturer/tradename

Cytiva 10600094

sheet W × L

80 mm × 90 mm

pore size

0.2 μm pore size

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF16935396GF89164873GF83929674
assay

95%

assay

95%

assay

≥95%

assay

95%

manufacturer/tradename

Goodfellow 837-700-94

manufacturer/tradename

Goodfellow 169-353-96

manufacturer/tradename

Goodfellow 891-648-73

manufacturer/tradename

Goodfellow 839-296-74

mp

630 °C (lit.)

mp

630 °C (lit.)

mp

630 °C (lit.)

mp

630 °C (lit.)

density

6.69 g/mL at 25 °C (lit.)

density

6.69 g/mL at 25 °C (lit.)

density

6.69 g/mL at 25 °C (lit.)

density

6.69 g/mL at 25 °C (lit.)

bp

1635 °C (lit.)

bp

1635 °C (lit.)

bp

1635 °C (lit.)

bp

1635 °C (lit.)

General description

Nitrocellulose membranes with a 0.2 µm pore size, low background, and excellent signal-to-noise ratios for Western blot protocols. Amersham Protran nitrocellulose membranes offer excellent sensitivity, resolution, and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western blotting detection reagents.

Application

Amersham Protran® Western blotting membranes, nitrocellulose is suitable for use in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • Low background and excellent signal-to-noise ratios, without the need for stringent washing conditions systems.
  • Compatible with a variety of Western blot labeling and detection systems
  • Risk of “blow-through” reduced by high binding capacity of small proteins (< Mr 20 000)
  • Optimized for chemiluminescent and fluorescent detection
  • Long shelf life of bound proteins to the transfer membrane
  • Available in popular precut sizes for Western blot transfer applications

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service