GE10600048
Amersham™ Protran® Premium Western blotting membranes, nitrocellulose
pore size 0.45 μm, sheet W × L 200 mm × 200 mm, pkg of 10 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
Recommended Products
packaging
pkg of 10 ea
manufacturer/tradename
Cytiva 10600048
sheet W × L
200 mm × 200 mm
pore size
0.45 μm pore size
Looking for similar products? Visit Product Comparison Guide
General description
Nitrocellulose blotting membrane, provides excellent sensitivity, resolution, and low background in fluorescence Western blotting detection applications.
Application
Amersham™ Protran® Premium Western blotting membranes, nitrocellulose may be used in:
- Western blotting
- Southern blotting
- Northern blotting
Features and Benefits
- High binding capacity (162 – 180 µg IgG/cm2).
- Suitable for proteins of a wide range of molecular weights.
- Unsupported, 100% pure nitrocellulose membrane for use with all ECL Western blotting systems and protein blotting applications.
- Available in popular precut sizes for laboratory application.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service