GBL724022
Grace Bio-Labs HybriSlip™ hybridization cover
L × W × thickness 22 mm × 40 mm × 0.25 mm, pkg of 1000 ea
Synonym(s):
coverslide, hydrophobic coverslip, nuclease free coverslip, thin plastic coverslip
All Photos(2)
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Recommended Products
material
colorless
polycarbonate
sterility
non-sterile
packaging
pkg of 1000 ea
manufacturer/tradename
Grace Bio-Labs 724022
L × W × thickness
22 mm × 40 mm × 0.25 mm
General description
HybriSlip™ ideally suited for in situ hybridization, in situ PCR and hybridization to genomic arrays on glass slides.
HybriSlips are flexible, thin (0.25 mm) covers that minimize fluid friction and facilitate uniform reagent distribution. RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Product is ready-to-use without pretreatment. Working surfaces are protected with clean release liners to prevent RNase contamination. HybriSlips remain flat and will not curl, even at high temperatures.
HybriSlips are flexible, thin (0.25 mm) covers that minimize fluid friction and facilitate uniform reagent distribution. RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Product is ready-to-use without pretreatment. Working surfaces are protected with clean release liners to prevent RNase contamination. HybriSlips remain flat and will not curl, even at high temperatures.
- RNase and DNase free
- Hydrophobic
- Disposable
- Will not chip or break
Legal Information
HybriSlip is a trademark of Grace Bio-Labs, Inc.