EP5825723007
Adapter for Eppendorf® Rotor A-4-81/S-4x500
for 80 - 120mL round-bottom tubes, used with rotor A-4-81/S-4x500
Synonym(s):
Centrifugation adapter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103911
NACRES:
NB.15
Recommended Products
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5825723007
compatibility
used with rotor A-4-81/S-4x500
General description
Adapter for 80 - 120mL round-bottom tubes, for rotor A-4-81/S-4x500, set with 2 pieces
Application
CENTRIFUGATION
Features and Benefits
for 1 round-bottom tube 80 – 120 mL, for Rotor A-4-81 and Rotor S-4x5002 pcs.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service