EP5703000010
Eppendorf® Centrifuge 5702 R G
without rotor, AC/DC input 230 V, 50 - 60 Hz
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
40161701
NACRES:
NB.81
Recommended Products
AC/DC input
230 V AC, Schuko plug
feature
CE compliant
refrigerated
without rotor
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 5703000010 (EU)
parameter
temp. range at -9-40 °C
3000 × g
4400 rpm
technique(s)
centrifugation: suitable
Looking for similar products? Visit Product Comparison Guide
General description
The 5702 family of low speed, general purpose centrifuges was especially designed to meet the requirement of low- to mid-throughput research and cell culture labs. They are compact in design so that one can fit comfortably on every lab bench, quiet in operation to benefit your work environment, and provide 6 different rotor options to accommodate any type of tube.
Quality and safety
- Centrifuge 5702 for standard applications
- Refrigerated Centrifuge 5702 R for heat sensitive samples
Quality and safety
- Stainless steel rotor chamber is rust-free and easy to clean
- Imbalance detection for maximum safety
- Key lock to prevent accidental parameter changes
- Aerosol-tight caps for safe centrifugation of hazardous samples optionally available
Features and Benefits
- Max. speed: 3,000 × g (4,400 rpm)
- Very compact footprint to fit on every lab bench
- Very quiet operation to benefit your environment
- Low access height for simple loading and unloading of samples
- SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
- “At set RPM” function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
- Key lock to prevent unintentional adjustments
- Stainless steel rotor chamber is rust-free and easy to clean
- Electronic imbalance detection for maximum safety
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service