EP3880001018
Eppendorf® IsoTherm-System starter set
for 1.5-2.0 mL tubes, pkg of 1 ea
All Photos(1)
About This Item
UNSPSC Code:
41102404
NACRES:
NC.21
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 3880001018
Features and Benefits
- Eppendorf® IsoTherm-System starter set, includes IsoRack (work rack), IsoSafe (insulating box), and two IsoPacks (cool packs).
- Ideal for cooling, transporting, and storing deep-frozen samples.
- Prevents enzymes from becoming inactive as well as for PCR sample preparation, sample transport and slow thawing of frozen samples.
- Dry incubation technology reduces contamination risk associated with classical icebath incubation for safe sample handling.
- Holds up to 24 μ tubes for broad flexibility.
- Two versions of IsoPack (cold pack) available: White maintains 0 °C for up to 6 hours. Blue maintains -21 °C for up to 3 hour.
- Racks are stackable, autoclavable and can be centrifuged in the MTP rotor.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG