Recommended Products
biological source
Mouse x Mouse Hybridoma
ImmunoDon
CAF1 mouse spleen
Myeloma
P3X63Ag8
Reactivity
Anti-CD4, human
technique(s)
cell culture | mammalian: suitable
isotype
IgG2b
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse P3X63Ag8 x Mouse CAF1 spleen
Cell Line Description
The antibody is directed against the CD4 antigen on human T cells of the T helper subset.
Immunogen
Human peripheral blood T lymphocytes
Culture Medium
IDMEM + 2mM glutamine + 20% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service