Key Documents
AXYVT38470ULRS
Pipette tips in 384-tip format, robotic
size 70 μL, pkg of 5x3840tips/cs, racked, sterile
Synonym(s):
pipet tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
sterility
sterile
manufacturer/tradename
Axygen Inc VT-384-70UL-R-S
Corning
packaging
pkg of 5x3840tips/cs
size
70 μL
General description
  • Axygen® automation pipette tips in 384-tip format are designed for Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST, and 16ST head.
  • Manufactured to stringent specifications including automated assembly to minimize contamination risks and ensure consistent performance and quality.
  • The clear polypropylene high-quality tips are ideal for sensitive biological applications.
  • RNase-/DNase-free and nonpyrogenic.
Other Notes
Browse all pipette tips
Legal Information
Agilent is a registered trademark of Agilent Technologies, Inc.
Axygen is a registered trademark of Corning, Inc.
Velocity11 VPrep is a registered trademark of Corning, Inc.