Skip to Content
Merck
All Photos(1)

Key Documents

74717

Sigma-Aldrich

Octaethylene glycol monohexadecyl ether

BioXtra, ≥98.0% (GC)

Synonym(s):

C16E8, Hexadecyl octaethylene glycol ether, Hexadecyloctaglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15(OCH2CH2)8OH
CAS Number:
Molecular Weight:
594.86
Beilstein:
6490568
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Quality Level

product line

BioXtra

Assay

≥98.0% (GC)

form

solid

mol wt

594.86 g/mol

technique(s)

protein purification: suitable

mp

43-45 °C

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C32H66O9/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17-34-19-21-36-23-25-38-27-29-40-31-32-41-30-28-39-26-24-37-22-20-35-18-16-33/h33H,2-32H2,1H3

InChI key

YAMTWWUZRPSEMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyosang Yoon et al.
Biosensors & bioelectronics, 117, 267-275 (2018-06-18)
Electroplating of nanoporous Pt (nPt) induces an extremely strong tensile stress, which results in the exfoliation of nPt on flexible polymer substrate despite plasma treatment to improve adhesion. Here, we overcame this challenge by modifying flexible stainless-steel, and developed wearable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service