Skip to Content
Merck
All Photos(1)

Key Documents

NIST1946

Lake Superior fish tissue

NIST® SRM® 1946

Synonym(s):

Environmental Fish Tissue Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 5 x 7-9 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

General description

This Standard Reference Material (SRM) is a frozen fish tissue homogenate that was prepared from lake trout (Salvelinus namaycush namaycush) collected near the Apostle Islands in Lake Superior (U.S./Canada), and is intended primarily for use in evaluating analytical methods for the determination of polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, polybrominated diphenyl ether (PBDE) congeners, perfluorooctanesulfonic acid (PFOS), fatty acids (including omega-3 fatty acids), extractable fat, methylmercury, total mercury, proximates, a-hexabromocyclododecane (a-HBCD), and selected trace elements in fish tissue and similar matrices.

A unit of SRM 1946 consists of five bottles, each containing approximately 7 g to 9 g (wet basis) of frozen tissue homogenate. For more information, please refer to the COA and SDS.

SRM 1946_cert

SRM 1946_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatty acids in Great Lakes lake trout and whitefish.
Pantazopoulos P, et al.
Journal of Great Lakes Research,, 39(1), 120-127 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service