Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of disk
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Certified for the analytes listed below.
Carbon (C), Manganese (Mn), Phosphorus (P), Sulfur (S), SIlicon (Si), Copper (Cu), Nickel (Ni), Chromium (Cr), Vanadium (V), Molybdenum (Mo)
Reference/Informational value is provided for the following analytes.
Aluminium (Al), Arsenic (As), Titanium (Ti), Iron (Fe)
See certificate for values and more details at nist.gov/SRM.
Carbon (C), Manganese (Mn), Phosphorus (P), Sulfur (S), SIlicon (Si), Copper (Cu), Nickel (Ni), Chromium (Cr), Vanadium (V), Molybdenum (Mo)
Reference/Informational value is provided for the following analytes.
Aluminium (Al), Arsenic (As), Titanium (Ti), Iron (Fe)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service