MISP00013
Microfil® Spare Protector Cap
For use with Microfil® filtration systems
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116129
NACRES:
NB.24
Recommended Products
Quality Level
packaging
pkg of 1 unit
manufacturer/tradename
Microfil®
input
sample type food(s)
sample type beverage(s)
application(s)
bioburden testing
clinical
environmental
food and beverages
industrial qc
water monitoring
compatibility
for use with Microfil® filtration systems
shipped in
ambient
Related Categories
General description
The spare protector cap is an additional spare part of the Microfil® filtration system. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. The spare protector cap is used only during transport to hold the frit in place.
Application
The spare protector cap is an additional accessory of the Microfil® filtration system used in general microbiological analysis of beer, bottled water, pharmaceutical products analysis, environmental analysis, bioburden testing, and water monitoring.
Legal Information
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service